VWR Chemicals, Base Materials for Dehydrated Media
Supplier: VWR Chemicals
VWRC26208.297EA 361 SGD
VWRC26208.297 VWRC84687.0500 VWRC84688.0500 VWRC84618.0500 VWRC84620.0500 VWRC84623.0500 VWRC84609.0500 VWRC84601.0500 VWRC84616.0500 VWRC84610.0500 VWRC84615.0500 VWRC84862.25000 VWRC84601.5000 VWRC84609.5000 VWRC84610.5000 VWRC84620.5000 VWRC84688.5000 VWRC85862.5000 VWRC85862.0500 VWRC84615.5000
VWR Chemicals, Base Materials for Dehydrated Media
Media for Microbiology Base Materials for Microbiology
The ingredients used to prepare culture media are very important, because ultimately the results depend on the quality of these compounds. Raw materials should be stored at 10 to 30 °C unless otherwise stated, in their closed containers in a dry environment.
  • Ingredients are specifically selected to ensure consistent, reproducible results
  • Manufactured under the strictest cGMP standards