VWR®, UV/Visible spectroscopy wavelength standards
Supplier: VWR Chemicals
VWRC84772.600EA 3030 SGD
VWRC84772.600 VWRC84773.600 VWRC84774.600 VWRC84775.180 VWRC84776.180 VWRC84777.180
VWR®, UV/Visible spectroscopy wavelength standards
Standards Colour Reference and Spectroscopy Standards
Wavelength standards (certified at 0,1; 0,2; 0,5; 1 and 2 nm slit widths) packed in six permanently sealed UV cuvettes or 100 ml amber bottles.
Order Now
SPECIFICATIONS
Environmentally Preferable