Chromium(III) chloride hexahydrate 98-101, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

Synonyms: Chromium trichloride hexahydrate

VWRC84839.260EA 120 SGD
VWRC84839.260
Chromium(III) chloride hexahydrate 98-101, GPR RECTAPUR®
Chromium (III) chloride hexahydrate
Formula: CrCl₃·6H₂O
MW: 266.45 g/mol
Melting Pt: 85
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149660
CAS Number: 10060-12-5
EINECS: 233-038-3
UN: 3260
ADR: 8 III
Merck Index: 13,02242

Order Now

Specification Test Results

Assay 98 - 101 %
Insolubility in water Max. 100 ppm
Substances not precipitated by NH4OH Max. 0.2 %
SO4 (Sulphate) Max. 100 ppm
Fe (Iron) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR