Lead(II) nitrate ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur.

Supplier: VWR Chemicals
Danger

Synonyms: Lead dinitrate

VWRC84847.180EA 31 SGD
VWRC84847.180 VWRC84847.290
Lead(II) nitrate ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur.
Lead (II) nitrate
Formula: Pb(NO₃)₂
MW: 331.21 g/mol
Melting Pt: 450…470 °C
Density: 4.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1 (6.1) II
Merck Index: 13,05432

Order Now

Specification Test Results

Assay Min. 99.0 %
Insoluble matter Max. 50 ppm
Cl (Chloride) Max. 10 ppm
Ca (Calcium) Max. 50 ppm
Cu (Copper) Max. 20 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 50 ppm
Na (Sodium) Max. 0.02 %
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR