Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS

Supplier: VWR Chemicals


Packaged under nitrogen.

VWRC84871.320EA 276 SGD
VWRC84871.320
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Water with 0,1% (v/v) trifluoroacetic acid
Storage Temperature: Ambient
EINECS: 200-929-3

Order Now

Specification Test Results

Nominal concentration (V/V) 0.095 - 0.105 %
Evaporation residue Max. 0.0001 %
Ca (Calcium) Max. 0.2 ppm
K (Potassium) Max. 0.2 ppm
Mg (Magnesium) Max. 0.2 ppm
Na (Sodium) Max. 0.5 ppm
Transmittance (210 nm) Min. 20 %
Transmittance (220 nm) Min. 50 %
Transmittance (230 nm) Min. 80 %
Transmittance (240 nm) Min. 95 %
Transmittance (250 nm) Min. 99 %
Suitable for LC-MS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR