Methanol with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS

Supplier: VWR Chemicals
Danger

VWRC84896.290EA 360 SGD
VWRC84896.290
Methanol with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Methanol with 0,1% (v/v) formic acid
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
UN: 1230
ADR: 3 (6.1),II

Order Now

Specification Test Results

Nominal concentration (V/V) 0.095 to 0.105 %
Evaporation residue Max. 0.0001 %
Water Max. 0.1 %
Ca (Calcium) Max. 0.2 ppm
K (Potassium) Max. 0.2 ppm
Mg (Magnesium) Max. 0.2 ppm
Na (Sodium) Max. 0.5 ppm
Suitable for LC-MS Passes test
Transmittance (240 nm) Min. 50 %
Transmittance (250 nm) Min. 80 %
Transmittance (260 nm) Min. 95 %
Transmittance (270 nm) Min. 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR