VWR®, Perchlorate 1,000 mg/l in Water standard for ion chromatography
Supplier: VWR Chemicals
VWRC84987.180EA 164 SGD
VWRC84987.180
VWR®, Perchlorate 1,000 mg/l in Water standard for ion chromatography
Standards Anionic Standards Perchlorate Standards
Certifications: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.