Potassium permanganate 0.002 mol/l (0.01 N) in aqueous solution

Supplier: VWR Chemicals

Synonyms: Permanganic acid potassium salt

VWRC85021.290EA 80.6 SGD
VWRC85021.290
Potassium permanganate 0.002 mol/l (0.01 N) in aqueous solution
Potassium permanganate
Formula: KMnO₄
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR