Copper(II) sulphate 0.5 mol/l USP Test Solution (TS)

Supplier: VWR Chemicals

Synonyms: Cupric sulphate

VWRC85343.180EA 105 SGD
VWRC85343.180 VWRC85343.260
Copper(II) sulphate 0.5 mol/l USP Test Solution (TS)
Copper (II) sulphate

Ready to use test solution (TS).


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Formula: CuSO₄
MW: 159.61 g/mol
Melting Pt: 200
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 0
UN: 3082
ADR: 9 III
Merck Index: 12,02722

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR