Iron(III) chloride in aqueous solution USP Test Solution (TS)

Supplier: VWR Chemicals

Synonyms: Iron trichloride, Ferric chloride

VWRC85347.180EA 139 SGD
VWRC85347.180
Iron(III) chloride in aqueous solution USP Test Solution (TS)
Iron (III) chloride

Ready to use test solution (TS).


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Formula: FeCl₃
Melting Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 0
UN: 2582
ADR: 8,III
Merck Index: 13,04048

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR