Oxalic acid 6,3% in aqueous solution USP Test Solution (TS)

Supplier: VWR Chemicals

Synonyms: Ethanedioic acid

VWRC85358.260EA 179 SGD
VWRC85358.260
Oxalic acid 6,3% in aqueous solution USP Test Solution (TS)
Oxalic acid

Ready to use test solution (TS).


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Formula: C₂H₂O₄
Melting Pt: 99 to 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 0
UN: 3265
ADR: 8,III
Merck Index: 13,06980

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR