Potassium permanganate 0.1 N in aqueous solution USP Test Solution (TS)

Supplier: VWR Chemicals

Synonyms: Permanganic acid potassium salt

VWRC85367.180EA 62.3 SGD
VWRC85367.180 VWRC85367.260
Potassium permanganate 0.1 N in aqueous solution USP Test Solution (TS)
Potassium permanganate

Ready to use test solution (TS).


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 0
UN: 3082
ADR: 9 III
Merck Index: 12,07824

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR