Sodium acetate in aqueous solution USP Test Solution (TS) 34

Supplier: VWR Chemicals

VWRC85371.260EA 237 SGD
VWRC85371.260
Sodium acetate in aqueous solution USP Test Solution (TS) 34
Sodium acetate

Ready to use test solutions (TS).


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Formula: H₃CCOONa
Melting Pt: 324 °C
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 0
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR