Ethyl acetate ≥99.9%, PESTINORM® Supra Trace for organic trace analysis

Supplier: VWR Chemicals
Danger

Synonyms: Acetic acid ethyl ester

Filtered through a 0.2 µm filter, packaged under inert gas.

VWRC85387.320EA 199 SGD
VWRC85387.320
Ethyl acetate ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.9 %
Evaporation residue Max 0.0003%
Colouration Max 10 APHA
Water Max 0.03%
GC/ECD Dioxins, Furans & PCB's Max 5 ng/l
GC/ECD any pesticide (as Lindane) Max 5 ng/l
GC/NPD any pesticide (as Parathion) Max 10 ng/l
GC/ECD 1,2,4-TCB to daca-PCB (as Lindane) Max 5 pg/ml
GC/ECD DCM to 1,2,4-TCB (as TCM) Max 2 ng/l
GC/FID C10 to C40 (as n-Decane) Max 2 ng/l
GC/MSD C10 to C40 (as Decane; 30-600amu) Max 2 ng/l
Filtered through 0.2um, filled under inert gas Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR