n-Hexane ≥95%, PESTINORM® Supra Trace for organic trace analysis

Supplier: VWR Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

Filtered through a 0.2 µm filter, packaged under inert gas.

VWRC85389.320EA 222 SGD
VWRC85389.320
n-Hexane ≥95%, PESTINORM® Supra Trace for organic trace analysis
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94,3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 95%
Evaporation residue Max 0.0003%
Colouration Max 10 APHA
Water Max 0.005%
GC/ECD Dioxins, Furans & PCB's Max 2 ng/l
GC/ECD any pesticide (as Lindane) Max 2 ng/l
GC/NPD any pesticide (as Parathion) Max 5 ng/l
GC/ECD 1,2,4-TCB to daca-PCB (as Lindane) Max 2 pg/ml
GC/ECD DCM to 1,2,4-TCB (as TCM) Max 1 ng/l
GC/FID C10 to C40 (as n-Decane) Max 2 ng/l
GC/MSD C10 to C40 (as Decane; 30-600amu) Max 2 ng/l
Filtered through 0.2um, filled under inert gas Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR