VWR® Rota® Tri, pH indicator, 1 to 11
Supplier: VWR Chemicals
VWRC85409.600EA 214 SGD
VWRC85409.600
VWR® Rota® Tri, pH indicator, 1 to 11
Test Papers pH Indicator Papers and Test Strips
These tri indicator papers show three different colours for each pH-value.
  • High precision due to the 3 colour scale
  • Easy to use
  • Produced from high quality paper to give good colour matching of paper to printed calibration scale
Tri paper has a hydrophobic barrier between the different indicators which ensures a distinctive separation of the colour bands.
Delivery information: Supplied in plactic reels that ensure protection and long term stability. Reel of 6 m length, width: 14 mm
Order Now
SPECIFICATIONS
Environmentally Preferable