VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
VWRC85410.601EA 33.9 SGD
VWRC85410.601 VWRC85411.601 VWRC85412.601 VWRC85413.601 VWRC85414.601 VWRC85415.601 VWRC85416.601 VWRC85417.601 VWRC85418.601 VWRC85419.601 VWRC85420.601 VWRC85421.601 VWRC85422.601 VWRC85423.601
VWR® Dosatest, pH indicator test strips, non bleeding
Test Papers pH Indicator Papers and Test Strips
Dosatest test strips provide precise and rapid pH determination, with up to four different colour blocks for each pH value.
  • Easier to work with due to the long handle
  • Colour-fixed reactive pads
  • Brilliant colour chart
pH indicator is chemically bound onto the cellulose fibre preventing bleeding. This effectively protects the sample against contamination and enables measurements to be taken even in weak buffered or strong alkaline solutions.
Better access to different types of containers preventing fingers coming into contact with the sample.
Order Now
SPECIFICATIONS
Environmentally Preferable