Test strips, peracetic acid, Dosatest
Supplier: VWR Chemicals
VWRC85437.601EA 86.1 SGD
VWRC85437.601 VWRC85436.601 VWRC85438.601
Test strips, peracetic acid, Dosatest
Environmental Test Kits
A ready to use kit that fits into your lab coat pocket.
  • Rapid and convenient
  • Easy to handle
  • Choice of ranges for required precision
Ideal to monitor your desinfection process. Semi-quantitative results can be obtained between 10 and 120 seconds
Colour charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Packaging: Container with 100 test strips 6×95 mm.