Ethyl acetate 99,9, HiPerSolv CHROMANORM® LC-MS

Supplier: VWR Chemicals
Danger

Synonyms: Acetic acid ethyl ester

Filter 0,2 um_Nitrogen_filled

VWRC85481.320EA 278 SGD
VWRC85481.320
Ethyl acetate 99,9, HiPerSolv CHROMANORM® LC-MS
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless liquid
Acidity (as Acetic acid) max. 0.003 %
Alkalinity (as Ammonia) max. 0.0005 %
Color (APHA, Pt-Co) max. 10
Assay (GC, on anhydrous basis) min. 99.9 %
Residue after evaporation max. 0.0005 %
Water (KF) max. 0.05 %
T260nm min. 75 %
T275nm min. 98 %
LC-MS suitability test Complies
Ag (Silver) max. 0.1 ppm
Al (Aluminum) max. 0.05 ppm
Ba (Barium) max. 0.1 ppm
Bi (Bismuth) max. 0.1 ppm
Ca (Calcium) max. 0.05 ppm
Cd (Cadmium) max. 0.05 ppm
Co (Cobalt) max. 0.05 ppm
Cr (Chromium) max. 0.02 ppm
Cu (Copper) max. 0.02 ppm
Fe (Iron) max. 0.02 ppm
K (Potassium) max. 0.05 ppm
Li (Lithium) max. 0.1 ppm
Mg (Magnesium) max. 0.05 ppm
Mn (Manganese) max. 0.02 ppm
Mo (Molybdenum) max. 0.05 ppm
Na (Sodium) max. 0.05 ppm
Ni (Nickel) max. 0.02 ppm
Pb (Lead) max. 0.02 ppm
Sn (Tin) max. 0.05 ppm
Sr (Strontium) max. 0.05 ppm
Zn (Zinc) max. 0.1 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR