Hydrochloric acid 29 - 31%, NORMATOM® for trace metal analysis

Supplier: VWR Chemicals
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

VWRC85493.290EA 261 SGD
VWRC85493.290
Hydrochloric acid 29 - 31%, NORMATOM® for trace metal analysis
Hydrochloric acid
Formula: HCl
MW: 36.46
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II
REACH: 01-2119484862-27

Order Now

Specification Test Results

Assay 29 - 31 %
Colouration max. 10 APHA
Br (Bromide) max. 10 ppm
Cl2 (Free Chlorine) max. 0.5 ppm
NH4 (Ammonium) max. 0.5 ppm
P (Total Phosphorus) max. 0.01 ppm
S (Total Sulphur) max. 0.3 ppm
Ag (Silver) max. 1 ppb
Al (Aluminium) max. 1 ppb
As (Arsenic) max. 0.5 ppb
Au (Gold) max. 0.5 ppb
B (Boron) max. 1 ppb
Ba (Barium) max. 0.1 ppb
Be (Beryllium) max. 0.1 ppb
Bi (Bismuth) max. 0.1 ppb
Ca (Calcium) max. 1 ppb
Cd (Cadmium) max. 0.1 ppb
Ce (Cerium) max. 0.1 ppb
Co (Cobalt) max. 0.1 ppb
Cr (Chromium) max. 0.5 ppb
Cs (Cesium) max. 0.1 ppb
Cu (Copper) max. 0.5 ppb
Dy (Dysprosium) max. 0.1 ppb
Er (Erbium) max. 0.1 ppb
Eu (Europium) max. 0.1 ppb
Fe (Iron) max. 1 ppb
Ga (Gallium) max. 0.1 ppb
Gd (Gadolinium) max. 0.1 ppb
Ge (Germanium) max. 1 ppb
Hf (Hafnium) max. 0.1 ppb
Hg (Mercury) max. 0.1 ppb
Ho (Holmium) max. 0.1 ppb
In (Indium) max. 0.1 ppb
K (Potassium) max. 1 ppb
La (Lanthanum) max. 0.1 ppb
Li (Lithium) max. 0.1 ppb
Lu (Lutetium) max. 0.1 ppb
Mg (Magnesium) max. 0.5 ppb
Mn (Manganese) max. 0.1 ppb
Mo (Molybdenum) max. 0.1 ppb
Na (Sodium) max. 1 ppb
Nb (Niobium) max. 0.1 ppb
Nd (Neodymium) max. 0.1 ppb
Ni (Nickel) max. 0.5 ppb
Pb (Lead) max. 0.1 ppb
Pd (Palladium) Information Only
Pr (Praseodymium) max. 0.1 ppb
Pt (Platinum) Information Only
Rb (Rubidium) max. 0.1 ppb
Re (Rhenium) max. 0.1 ppb
Rh (Rhodium) max. 0.1 ppb
Ru (Ruthenium) max. 0.1 ppb
Sb (Antimony) max. 0.5 ppb
Sc (Scandium) max. 0.1 ppb
Se (Selenium) max. 1 ppb
Sm (Samarium) max. 0.1 ppb
Sn (Tin) max. 0.5 ppb
Sr (Strontium) max. 0.1 ppb
Tb (Terbium) max. 0.1 ppb
Te (Tellurium) max. 0.1 ppb
Th (Thorium) max. 0.1 ppb
Ti (Titanium) max. 0.5 ppb
TI (Thallium) max. 0.1 ppb
Tm (Thulium) max. 0.1 ppb
U (Uranium) max. 0.1 ppb
V (Vanadium) max. 0.5 ppb
W (Tungsten) max. 0.1 ppb
Y (Yttrium) max. 0.1 ppb
Yb (Ytterbium) max. 0.1 ppb
Zn (Zinc) max. 1 ppb
Zr (Zirconium) max. 0.1 ppb

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR