Formic acid 90% ≥90%, AnalaR NORMAPUR® ACS

Supplier: VWR Chemicals
Danger

Synonyms: Aminic Acid, Methanoic acid

VWRC85495.290EA 166 SGD
VWRC85495.290
Formic acid 90% ≥90%, AnalaR NORMAPUR® ACS
Formic acid
Formula: HCO₂H
Density: 1,208 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 0
UN: 1779
ADR: 8 (3),II
Merck Index: 13,04265
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 89.0 - 91.0 %
Dilution test Passes test ACS
SO3 (Sulphite) Passes test
Colouration Max. 10 APHA
Acetic acid Max. 0.04 %
Evaporation residue Max. 20 ppm
Heavy metals (by ICP-MS) Max. 5 ppm
Cl (Chloride) Max. 10 ppm
NH4 (Ammonium) Max. 50 ppm
SO4 (Sulphate) Max. 20 ppm
Fe (Iron) Max. 5 ppm
Conforms to ACS Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR