Barium chloride 0.1 mol/l (0.2N) in aqueous solution, AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

VWRC85496.230EA 88.3 SGD
VWRC85496.230
Barium chloride 0.1 mol/l (0.2N) in aqueous solution, AVS TITRINORM volumetric solution
Barium chloride
Formula: BaCl₂
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0,0998 -> 0,1002 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR