Ammonium sulphate 99,0 Reag. Ph. Eur.

Supplier: VWR Chemicals

Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

VWRC85509.290EA 41.7 SGD
VWRC85509.290
Ammonium sulphate 99,0 Reag. Ph. Eur.
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: 336…339 °C
Density: 1.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Assay Min. 99.0 %
Appearance White crystals
pH (5 %) 4.5 - 6.0
Sulphated ash Max. 0.1 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR