VWR®, Powder packs for determination of aluminium, reagents for photometers
Supplier: VWR Chemicals
VWRC85600.620EA 394 SGD
VWRC85600.620
VWR®, Powder packs for determination of aluminium, reagents for photometers
Environmental Test Kits
VWR® powder pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary. The pillows, individually sealed in aluminium foil packs, are unaffected by ambient conditions and contain pre-measured powder reagents for accuracy, convenience and great solubility.
  • Accurate
  • Fast and convenient by easier dissolution than tablets and long shelf life
  • No calibration necessary
  • Beveled edge prevents reagent from getting stuck in corners
RS = Powder Pack reagent set