VWR®, Powder packs for determination of sulphate, reagents for photometers
Supplier: VWR Chemicals
VWRC85628.620EA 112 SGD
VWRC85628.620 VWRC85628.680 VWRC85629.620
VWR®, Powder packs for determination of sulphate, reagents for photometers
Environmental Test Kits
VWR® Powder Pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary. The pillows, individually sealed in aluminium foil packs, are unaffected by ambient conditions and contain pre-measured powder reagents for accuracy, convenience and great solubility.
  • Accurate
  • Fast and convenient by easier dissolution than tablets and long shelf life
  • No calibration necessary
  • Beveled edge prevents reagent from getting stuck in corners
Order Now
SPECIFICATIONS
Environmentally Preferable