VWR®, Uranium standard solutions for ICP-MS, ARISTAR®
Supplier: VWR Chemicals
VWRC85641.180EA 200 SGD
VWRC85641.180
VWR®, Uranium standard solutions for ICP-MS, ARISTAR®
Standards Cationic Standards Uranium Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete certificate of analysis
Certifications: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities