VWR® Conductivity standard, CRM, 5000 μS/cm
Supplier: VWR Chemicals
VWRC85645.260EA 304 SGD
VWRC85645.260
VWR® Conductivity standard, CRM, 5000 μS/cm
Standards Conductivity Standards
Directly traceable to NIST, tested and certified to ISO17025 and ISO 17034