VWR® PESTINORM®, Ethanol, Secondary Reference Standard for GC
Supplier: VWR Chemicals
VWRC85665.180EA 207 SGD
VWRC85665.180
VWR® PESTINORM®, Ethanol, Secondary Reference Standard for GC
Standards Chromatography Standards
A secondary GC solvent reference standard for various chromatographic and analytical applications.
  • Purity and the specific impurity profile is determined by GC-MS and reported in the CoA
  • Traceability data to specific current lot of primary standard
  • Completely synthetic and typically over 99,9% pure by GC-FID