Total Hardness Test Strips, Dosatest, VWR®
Supplier: VWR Chemicals
VWRC85694.610EA 683 SGD
VWRC85694.610
Total Hardness Test Strips, Dosatest, VWR®
Environmental Test Kits
Semi-quantitative test strips for the determination of water hardness.
  • Ideal for rapid and easy analysis of water hardness directly on-site
  • No maintenance or calibration is required
Gradation:
- Total hardness <3.75 • >5 • >8.75 • >17.5 • >26.25 °e
- Total hardness <54 • >72 • >126 • >252 • >378 ppm CaCO₃
Color change: Total hardness - green → red
Packaging:
Box of 1000 loose strips, individually sealed
Caution: Each test field on the test strips contains mercury(II) bromide. Toxic in contact with skin