Chromate test strips, Dosatest
Supplier: VWR Chemicals
VWRC85695.601EA 95.3 SGD
VWRC85695.601
Chromate test strips, Dosatest
Environmental Test Kits
Semi-quantitative test strips for the determination of chromate.
  • Ideal for rapid and easy analysis of chromate directly on-site
  • No maintenance or calibration is required
Colour change: White to violet.
Delivery information: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box