VWR®, Zinc Test Strips, Dosatest
Supplier: VWR Chemicals
VWRC85700.601EA 122 SGD
VWRC85700.601
VWR®, Zinc Test Strips, Dosatest
Environmental Test Kits
Semi-quantitative test strips for the determination of zinc.
  • Ideal for rapid and easy analysis of zinc directly on-site
  • No maintenance or calibration is required
Colour change: Orange to red
Delivery information: Delivered as 100 test strips in a tube with reagents measuring vessel and instruction leaflet.
Caution: Strongly acid solutions (pH < 2) have to be buffered to a pH value of 2 to 6 using solid sodium acetate.