Ascorbic acid test strips, Dosatest
Supplier: VWR Chemicals
VWRC85701.601EA 65.9 SGD
VWRC85701.601
Ascorbic acid test strips, Dosatest
Environmental Test Kits
Semi-quantitative test strips for the determination of ascorbic acid.
  • Ideal for rapid and easy analysis directly on-site
  • No maintenance or calibration is required
Colour change: Yellow to green-blue
Delivery information: Delivered as 100 test strips in a tube with instruction leaflet.