Nitrite test strips, Dosatest
Supplier: VWR Chemicals
VWRC85705.601EA 70 SGD
VWRC85705.601
Nitrite test strips, Dosatest
Environmental Test Kits
Semi-quantitative test strips for the determination of nitrite.
  • Ideal for rapid and easy analysis of nitrite directly on-site
  • No maintenance or calibration is required
Colour change: Yellow to red
Delivery information: Delivered as 100 test strips in a tube, instruction leaflet in a box
Caution: Strongly acidic solutions must be diluted with dilute sodium hydroxide and strongly alkaline samples with dilute sulphuric acid to pH 4 to 6.