Sulphate test strips, Dosatest, VWR®
Supplier: VWR Chemicals
VWRC85708.601EA 62.6 SGD
VWRC85708.601
Sulphate test strips, Dosatest, VWR®
Environmental Test Kits
Semi-quantitative test strips for the determination of sulphate.
  • Ideal for rapid and easy analysis of sulphate directly on-site
  • No maintenance or calibration is required
Colour change: Red to yellow
Delivery information: Delivered as 100 test strips in a tube with instruction leaflet.
Caution: If the sample solution has a pH value of 4 to 8, the reaction will take place without interferences. Strongly acid solutions have to be buffered with sodium acetate, and alkaline with tartaric acid.
Order Now
SPECIFICATIONS
Environmentally Preferable