Arsenic test strips, Dosatest
Supplier: VWR Chemicals
VWRC85710.601EA 217 SGD
VWRC85710.601
Arsenic test strips, Dosatest
Environmental Test Kits
Semi-quantitative test strips for the determination of arsenic.
  • Ideal for rapid and easy analysis of arsenic directly on-site
  • No maintenance or calibration is required
Colour change: White to yellow-brown
Delivery information: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box
Caution: Each test field on the test strips contains mercury(II) bromide. Toxic in contact with skin