VWR® AVS TITRINORM, Cesium chloride/aluminium nitrate buffer solution acc. to schuhknecht and schinkel for determination of na, k and li by flame aas
Supplier: VWR Chemicals
VWRC85772.290EA 441 SGD
VWRC85772.290
VWR® AVS TITRINORM, Cesium chloride/aluminium nitrate buffer solution acc. to schuhknecht and schinkel for determination of na, k and li by flame aas
Buffers Ionisation Buffers
This material is intended for use as an ionisation buffer for AAS
Content of aluminium nitrate nonahydrate (Al(NO₃)₃.9H2O) – 250 g/L
Content of cesium chloride (CsCl) – 50 g/L
Impurities
- K (Potassium) < 1.0 mg/l
- Li (Lithium) < 1.0 mg/l
- Na (Sodium) < 5.0 mg/l