Magnesium nitrate 10 g/L (99,999%) in water, ARISTAR® matrix modifier solution, for graphite furnace AAS

Supplier: VWR Chemicals

VWRC85780.154EA 281 SGD
VWRC85780.154
Magnesium nitrate 10 g/L (99,999%) in water, ARISTAR® matrix modifier solution, for graphite furnace AAS
Magnesium nitrate solutions

The VWR® Matrix modifier solutions have especially been developed for use in GF-AAS to eliminate matrix effects. They can be used either diluted or as mixtures.


  • Guaranteed accuracy: ±1%
  • COA incl. up to 70 measured trace impurities in ppm range


VWR® Matrix modifier solutions are produced with high purity metals or salts (≥99,999%) in sub-boiling distilled acids and are supplied with a detailed batch specific certificate of analysis.

Formula: Mg(NO₃)₂
Storage Temperature: Ambient
CAS Number: 13446-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR