1-Butanesulphonic acid sodium salt ≥99.0% (dried basis), HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals

Synonyms: Sodium 1-butanesulphonate, Sodium butane-1-sulphonate, Sodium 1-butansulphonate, Butane-1-sulphonic acid sodium salt

VWRC85790.150EA 97.2 SGD
VWRC85790.150
1-Butanesulphonic acid sodium salt ≥99.0% (dried basis), HiPerSolv CHROMANORM® for HPLC
1-Butansulphonic acid sodium salt
Formula: C₄H₉NaO₃S
MW: 160.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007540
CAS Number: 2386-54-1

Order Now

Specification Test Results

Assay (calculated on dried substance) ≥ 99,0 %
Appearance White powder
IR Spectrum Passes test
Appearance of solution (10 %) Clear and colourless
Water ≤ 1,0 %
Absorbance (210 nm) (0.5 mol/l) ≤ 0,1
Absorbance (220 nm) (0.5 mol/l) ≤ 0,06
Absorbance (230 nm) (0.5 mol/l) ≤ 0,04
Absorbance (260 nm) (0.5 mol/l) ≤ 0,02


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR