Sodium-1-decanesulphonate ≥99.0%, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals

Synonyms: Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt

VWRC85792.150EA 97.3 SGD
VWRC85792.150
Sodium-1-decanesulphonate ≥99.0%, HiPerSolv CHROMANORM® for HPLC
Sodium 1-decanesulphonate
Formula: C₁₀H₂₁NaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00007526
CAS Number: 13419-61-9

Order Now

Specification Test Results

Assay (calculated on dried substance) ≥ 99,00 %
Appearance White to cream powder
IR Spectrum Passes test
Appearance of solution (5 %) Clear and colourless
Water ≤ 1,0 %
Absorbance (210 nm) (0.2 mol/l) ≤ 0,05
Absorbance (220 nm) (0.2 mol/l) ≤ 0,03
Absorbance (230 nm) (0.2 mol/l) ≤ 0,02
Absorbance (260 nm) (0.2 mol/l) ≤ 0,02


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR