Lithium dodecyl sulphate 98,5, HiPerSolv CHROMANORM® HPLC

Supplier: VWR Chemicals
Danger

Synonyms: Dodecyl sulphuric acid lithium salt, Dodecyl sulphate lithium salt, Lithium lauryl sulphate, Lauryl sulphate lithium salt

VWRC85793.150EA 97 SGD
VWRC85793.150
Lithium dodecyl sulphate 98,5, HiPerSolv CHROMANORM® HPLC
Lithium dodecyl sulphate
Formula: C₁₂H₂₅LiO₄S
MW: 272.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007467
CAS Number: 2044-56-6
UN: 1325
ADR: 4.1 III

Order Now

Specification Test Results

Assay (C12) ≥ 98,50 %
Appearance White fine crystals
IR Spectrum Passes test
Loss on drying (120°C; under vacuum) ≤ 0,50 %
Appearance of solution (1 %; ethanol) Clear and colourless
Appearance of solution (3 %) Clear and colourless
Cl (Chloride) ≤ 100,0 ppm
Absorbance (260 nm) (3 %) ≤ 0,100
Absorbance (280 nm) (3 %) ≤ 0,075

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR