Tetrabutylammonium hydrogen sulphate, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Warning

Synonyms: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulphate

VWRC85794.150EA 421 SGD
VWRC85794.150
Tetrabutylammonium hydrogen sulphate, HiPerSolv CHROMANORM® for HPLC
Tetrabutylammonium hydrogen sulphate
Formula: (C₄H₉)₄NHSO₄
Melting Pt: 169…172 °C
Density: 1,01 g/cm³ (25 °C)
MDL Number: MFCD00011637
CAS Number: 32503-27-8
EINECS: 251-068-5

Order Now

Specification Test Results

Assay (Acidimetric) ≥ 99,00 %
IR Spectrum Passes test
Absorbance (200 nm) (0.005 mol/l) ≤ 0,155
Absorbance (220 nm) (0.005 mol/l) ≤ 0,046
Absorbance (250 nm) (0.005 mol/l) ≤ 0,009


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR