n-Hexane 99, HiPerSolv CHROMANORM® LC-MS

Supplier: VWR Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

Nitrogen_filled

VWRC85799.320EA 520 SGD
VWRC85799.320 VWRC85799.290
n-Hexane 99, HiPerSolv CHROMANORM® LC-MS
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94,3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: -22
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
UN: 1208
ADR: 3
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on anhydrous) ≥ 99 %
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Evaporation residue ≤ 1 ppm
Water ≤ 50 ppm
Ag (Silver) ≤ 10 ppb
Al (Aluminium) ≤ 25 ppb
Ca (Calcium) ≤ 25 ppb
Cu (Copper) ≤ 10 ppb
Fe (Iron) ≤ 10 ppb
K (Potassium) ≤ 10 ppb
Mg (Magnesium) ≤ 10 ppb
Mn (Manganese) ≤ 10 ppb
Na (Sodium) ≤ 50 ppb
Ni (Nickel) ≤ 10 ppb
Pb (Lead) ≤ 10 ppb
Zn (Zinc) ≤ 10 ppb
Transmittance (190 nm) ≥ 10 %
Transmittance (205 nm) ≥ 50 %
Transmittance (220 nm) ≥ 80 %
Transmittance (235 nm) ≥ 95 %
Transmittance (255 nm) ≥ 99 %
Packaged under nitrogen Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR