Methanol ≥99.9%, HiPerSolv CHROMANORM® for UHPLC-MS

Supplier: VWR Chemicals
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

Filtered through a 0.1 micron filter.

VWRC85802.290EA 322 SGD
VWRC85802.290
Methanol ≥99.9%, HiPerSolv CHROMANORM® for UHPLC-MS
Methanol
Formula: CH₃OH
Boiling Pt: 64,6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0,7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1),II
REACH: 01-2119433307-44
Merck Index: 13,05984

Order Now

Specification Test Results

Assay (GC) min. 99.9%
Identity (IR) complying
Clear colorless liquid complying
Color (APHA Pt-Co) max. 10
Water(KF) max. 0.01% w/w
Residue after evaporation max. 0.7 ppm
Acidity max. 0.001 meq/g
Alkalinity max. 0.0002 meq/g
Aluminium (Al) max. 5 ppb
Calcium (Ca) max. 5 ppb
Iron (Fe) max. 5 ppb
Potassium (K) max. 5 ppb
Magnesium (Mg) max. 5 ppb
Sodium (Na) max. 25 ppb
Fluorescence 254/450 nm (as Quinine) max. 0.5 ppb
Fluorescence 365/450 nm (as Quinine) max. 0.5 ppb
UHPLC/MS ESI/APCI+ (as Reserpine) max. 2 ppb
UHPLC/MS ESI/APCI- (as Reserpine) max. 10 ppb
PEG Signal-to-noise ratio max. 50
UHPLC-UV Gradient (230 nm) max. 2.0 mAU
UHPLC-UV Gradient (254 nm) max. 0.5 mAU
UV Absorbance 210nm max. 0.50 AU
UV Absorbance 215 nm max. 0.40 AU
UV Absorbance 220 nm max. 0.20 AU
UV Absorbance 230 nm max. 0.10 AU
UV Absorbance 254 nm max. 0.01 AU
UV Absorbance 260 nm max. 0.005 AU
UV Absorbance 280 nm max. 0.005 AU
Filtered through 0.1 µm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR