Cerium(IV) sulphate 0.25 mol/l (0.25 N) in 0.25 N sulphuric acid, AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

Synonyms: Cerium disulphate, Ceric sulphate, Cerium(IV) sulphate

VWRC85804.290EA 688 SGD
VWRC85804.290
Cerium(IV) sulphate 0.25 mol/l (0.25 N) in 0.25 N sulphuric acid, AVS TITRINORM volumetric solution
Cerium (IV) sulphate
Formula: Ce(SO₄)₂
Storage Temperature: Ambient
MDL Number: MFCD00148852
CAS Number: 13590-82-4

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.2495 -> 0.2505 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR