Oxalic acid 0.1 mol/l (0.2 N) in aqueous solution, AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

Synonyms: Ethanedioic acid

VWRC85807.290EA 64.8 SGD
VWRC85807.290
Oxalic acid 0.1 mol/l (0.2 N) in aqueous solution, AVS TITRINORM volumetric solution
Oxalic acid
Formula: C₂H₂O₄
MDL Number: MFCD00002573
CAS Number: 144-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR