Potassium bromide 0.1 M in aqueous solution

Supplier: VWR Chemicals

VWRC85808.290EA 66.5 SGD
VWRC85808.290
Potassium bromide 0.1 M in aqueous solution
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
MDL Number: MFCD00011358
CAS Number: 7758-02-3
Merck Index: 13,07701

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR