Potassium chloride 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

Synonyms: Potassiumchloride

VWRC85809.290EA 79.5 SGD
VWRC85809.290
Potassium chloride 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Potassium chloride
Formula: KCl
MDL Number: MFCD00011360
CAS Number: 7447-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR