Potassium permanganate 0.01 mol/l (0.05 N) aqueous solution, AVS TITRINORM Volumetric

Supplier: VWR Chemicals

Synonyms: Permanganic acid potassium salt

VWRC85810.290EA 100 SGD
VWRC85810.290
Potassium permanganate 0.01 mol/l (0.05 N) aqueous solution, AVS TITRINORM Volumetric
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
UN: 3082
ADR: 9 III
Merck Index: 12,07824

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR