Sodium chloride 0,5 mol/l 0,5 N in aqueous solution, AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

VWRC85812.290EA 65 SGD
VWRC85812.290
Sodium chloride 0,5 mol/l 0,5 N in aqueous solution, AVS TITRINORM volumetric solution
Sodium chloride
Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR