Chlorophenol red, AnalaR NORMAPUR® Indicator

Supplier: VWR Chemicals

Synonyms: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide

VWRC85921.100EA 41.9 SGD
VWRC85921.100 VWRC85921.130
Chlorophenol red, AnalaR NORMAPUR® Indicator
Chlorophenol red
Formula: C₁₉H₁₂Cl₂O₅S
MW: 423.27 g/mol
Melting Pt: ∼261 °C
Storage Temperature: Ambient
MDL Number: MFCD00005877
CAS Number: 4430-20-0
EINECS: 224-619-2

Order Now

Specification Test Results

Green to red-brown to brown powder Passes test
Solubility in ethanol (0.1 %) Clear solution
Visual transition interval pH 4.8 - pH 6.4
Loss on drying ≤ 8.0 %
Absorption max. λ (0.1 mol/l NaOH) 572 - 575 nm
Absorptivity (1 %; 1 cm; λmax) ≥ 1000


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR